missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dysprosium ICP Standard, 1mL = 1mg Dy (1,000ppm Dy), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPDY1KN500

 View more versions of this product

Catalog No. PDY1KN-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Dysprosium ICP Standard
93.0,4.63,0.11
Liquid
1,000ppm Dy
Colorless
1308-87-8,7697-37-2,1308-87-8
97.0,4.81,0.11
Poly natural
1 mL = 1 mg Dy
500mL

Chemical Identifiers

1308-87-8
Documents