missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dysprosium Foil, 0.1mm (0.004in) thick, 0.999, Alfa Aesar™

Form: Approx. 0.53g/25 x 25mm

Manufacturer:  Thermo Fisher Scientific 12382FF

 View more versions of this product

Catalog No. AA12382FF



Specifications

Specifications

Dysprosium
99.90%
MFCD00010982
atom
[Dy]
162.5
CHEBI:33377
Odorless
25 x 25mm
7429-91-6
Dy
Gray to silver foil
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
Foil, 0.1mm (0.004 in.) thick
Gray to Silver

Chemical Identifiers

7429-91-6
162.5
KBQHZAAAGSGFKK-UHFFFAOYSA-N
23912
dysprosium
Dy
MFCD00010982
atom
CHEBI:33377
[Dy]
Safety and Handling

Safety and Handling

H260-H228; In contact with water releases flammable gases which may ignite spontaneously. Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-073-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents