missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dysprosium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals
Form: Approx. 0.13g/25 x 25mm
Supplier: Thermo Scientific Chemicals 012383FF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dysprosium | |
7429-91-6 | |
Foil | |
99.90% | |
MFCD00010982 | |
KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
dysprosium | |
23912 | |
Odorless | |
Gray to silver foil |
Rare Earth Oxides Content | |
Gray to Silver | |
25 x 25 mm | |
Dy | |
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
[Dy] | |
162.50 | |
CHEBI:33377 | |
Under Argon | |
8.55g/cm3 |
Chemical Identifiers
7429-91-6 | |
162.50 | |
KBQHZAAAGSGFKK-UHFFFAOYSA-N | |
23912 | |
dysprosium |
Dy | |
MFCD00010982 | |
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n | |
CHEBI:33377 | |
[Dy] |
Safety and Handling
Keep container tightly closed and dry.
H260-H228; In contact with water releases flammable gases which may ignite spontaneously. Flammable solid.
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-073-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only