missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dysprosium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

Form: Approx. 0.13g/25 x 25mm

$310.25 - $1165.92

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA12383FF
View Documents Promotion Details
Thermo Scientific Chemicals
012383FF
25 x 25 mm
Each for $310.25
 
AA12383FI
View Documents Promotion Details
Thermo Scientific Chemicals
012383FI
50 x 50 mm
Each for $1,165.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-91-6
162.50
KBQHZAAAGSGFKK-UHFFFAOYSA-N
23912
dysprosium
Dy
MFCD00010982
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
CHEBI:33377
[Dy]

Specifications

7429-91-6
Foil
99.90%
MFCD00010982
KBQHZAAAGSGFKK-UHFFFAOYSA-N
dysprosium
23912
Odorless
Gray to silver foil
Dysprosium, Rare Earth Oxides Content
Gray to Silver
25 x 25 mm
Dy
ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
[Dy]
162.50
CHEBI:33377
Under Argon
8.55g/cm3
Safety and Handling

Safety and Handling

Keep container tightly closed and dry.

H260-H228; In contact with water releases flammable gases which may ignite spontaneously. Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-073-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only