Invitrogen™ DYRK1B Recombinant Human Protein DFS Item
DYRK1B is a member of the DYRK family of protein kinases.
Manufacturer:  Invitrogen™ LSPV4650
 View more versions of this product
Catalog No. PV4650
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
DYRK1B is proposed to be involved in nuclear functions.
Specifications
Specifications
NP_004705
Frozen
9149
1mg
RUO - research use only
DYRK1B, MIRK
Baculovirus
Human
GST Tag
Target & Lead Identification & Validation|Kinase Biology|Kinase Proteins & Expression Reagents|Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis|Industrial & Applied Science|Pharma & Biopharma
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100
96.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
DYRK Kinase Family
Yes
Documents