DYRK1A Recombinant Human Protein DFS Item
DYRK1A Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3997
 View more versions of this product
Catalog No. PV3997
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
DYRK1A is a dual specificity kinase involved in cell proliferation. The gene for DYRK1A is located in the Down syndrome critical region of chromosome 21, and has been implicated in the pathogenesis of this disease.
Specifications
Specifications
NP_001387
Protein assays and analysis
Frozen
DYRK1A Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
DYRK1A, DYRK1
DYRK1A was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL DYRKtide peptide substrate (RRRFRPASPLRGPPK) and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
DYRK1A Recombinant Human Protein
Human
0.16mg/mL
Frozen
113kDa
85% by SDS-PAGE
–80°C
1 Each
DYRK1A Recombinant Human Protein
Kinase assay
DYRK
2880nmoles of phosphate transferred to DYRKtide peptide substrate (RRRFRPASPLRGPPK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.83μg/mL
GST Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.