missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dyphylline

An adenosine receptor antagonist, phosphodiesterase inhibitor and vasodilator

$122.50 - $610.91

Chemical Identifiers

CAS 479-18-5
Molecular Formula C10H14N4O4
Molecular Weight (g/mol) 254.246
MDL Number MFCD00005759
InChI Key KSCFJBIXMNOVSH-UHFFFAOYSA-N
Synonym 7-2,3-dihydroxypropyl theophylline, aristophyllin, diphylline, diprofilline, diprophyllin, diprophylline, dyphylline, glyphylline, lufyllin, neothylline
PubChem CID 3182
ChEBI CHEBI:4728
IUPAC Name 7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6316722 View Documents Alfa Aesar™
J6316722
100g Each for $122.50
 
AAJ6316736 View Documents Alfa Aesar™
J6316736
500g Each for $332.92
 
AAJ63167A1 View Documents Alfa Aesar™
J63167A1
1kg Each for $610.91
 
Specifications

Chemical Identifiers

479-18-5
254.246
KSCFJBIXMNOVSH-UHFFFAOYSA-N
3182
7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
C10H14N4O4
MFCD00005759
7-2,3-dihydroxypropyl theophylline, aristophyllin, diphylline, diprofilline, diprophyllin, diprophylline, dyphylline, glyphylline, lufyllin, neothylline
CHEBI:4728
CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O

Specifications

White
100g
C10H14N4O4
284563
7-2,3-dihydroxypropyl theophylline, aristophyllin, diphylline, diprofilline, diprophyllin, diprophylline, dyphylline, glyphylline, lufyllin, neothylline
KSCFJBIXMNOVSH-UHFFFAOYSA-N
7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
3182
254.24
Dyphylline
160°C to 165°C
479-18-5
MFCD00005759
14,3479
Soluble in water
CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
254.246
CHEBI:4728
Powder
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 207-526-1

RTECSNumber : XH5100000

TSCA : Yes

Recommended Storage : Ambient temperatures