missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dyphylline

An adenosine receptor antagonist, phosphodiesterase inhibitor and vasodilator

Manufacturer:  Alfa Aesar™ J63167A1

 View more versions of this product

Catalog No. AAJ63167A1



Specifications

Specifications

Dyphylline
160°C to 165°C
479-18-5
MFCD00005759
14,3479
Soluble in water
CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
254.246
CHEBI:4728
Powder
White
1kg
C10H14N4O4
284563
7-2,3-dihydroxypropyl theophylline, aristophyllin, diphylline, diprofilline, diprophyllin, diprophylline, dyphylline, glyphylline, lufyllin, neothylline
KSCFJBIXMNOVSH-UHFFFAOYSA-N
7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
3182
254.24

Chemical Identifiers

479-18-5
254.246
KSCFJBIXMNOVSH-UHFFFAOYSA-N
3182
7-(2,3-dihydroxypropyl)-1,3-dimethylpurine-2,6-dione
C10H14N4O4
MFCD00005759
7-2,3-dihydroxypropyl theophylline, aristophyllin, diphylline, diprofilline, diprophyllin, diprophylline, dyphylline, glyphylline, lufyllin, neothylline
CHEBI:4728
CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CC(CO)O
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 207-526-1

RTECSNumber : XH5100000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents