Please login to your online account to display your discounted pricing

Dynole 34-2, Tocris™

Dynamin I inhibitor

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Dynole 34-2
Assay Percent Range >99%
Molecular Formula C25H36N4O
Formula Weight 408.58
Synonym 2-Cyano-3-[1-[3-(dimethylamino)propyl]-1H-indol-3-yl]-N-octyl-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

422250

 
r & d systems
422250
50mg Each for $976.15

422210

 
r & d systems
422210
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 408.58
Quantity 50mg
Chemical Name or Material Dynole 34-2
Assay Percent Range >99%
Molecular Formula C25H36N4O
CAS 1128165-88-7
Synonym 2-Cyano-3-[1-[3-(dimethylamino)propyl]-1H-indol-3-yl]-N-octyl-2-propenamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C