Please login to your online account to display your discounted pricing

Dynein Mouse, Clone: 74.1, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $160.95

Specifications

Antigen Dynein
Clone 74.1
Host Species Mouse
Species Reactivity Bovine, Drosophila, Human, Mouse, Pig, Rat, Sheep, Xenopus
Applications Enzyme Linked Immunosorbent Assay
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122855

 
life technologies
LS14977280
25μg Each for $160.95
Description & Specifications

Specifications

Antigen Dynein
Clone 74.1
Host Species Mouse
Species Reactivity Bovine, Drosophila, Human, Mouse, Pig, Rat, Sheep, Xenopus
Applications Enzyme Linked Immunosorbent Assay
Applications Flow Cytometry (Intracellular Staining)
Applications Immunocytochemistry
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 74.1 recognizes all known forms of the 74kDa intermediate chain subunit of mammalian cytoplasmic dynein. Cytoplasmic dynein is a microtubule-stimulated ATPase responsible for movement of membrane-bound organelles in a directed manner toward the minus end of microtubules, commonly found at the centrosome. Dynein is responsible for the transport of lysosomes, endosomes, phagosomes, peroxisomes, mitochondria, and vesicles from the endoplasmic reticulum destined for the Golgi. During mitosis, dynein is responsible for positioning the spindle apparatus as well as being localized to the kinetochores to facilitate chromosome movement.

The 74.1 antibody recognizes human, mouse, rat, bovine, sheep, Xenopus, porcine, and Drosophila dynein intermediate chain.