Please login to your online account to display your discounted pricing
Thermo Scientific™ DyNAzyme Buffers
Choose from several formats of these buffers to optimize PCR using Thermo Scientific™ DyNAzyme™ DNA Polymerases.
Manufacturer: thermo scientific™  F517L
 View more versions of this product
Catalog No. F517L
  • / Each
Description & Specifications
Specifications
For Use With (Application) PCR, long PCR, DHPLC, microarray
Includes Detergent-free 10x Optimized DyNAzyme EXT Buffer
Quantity 4 x 1.5mL
Optimized Thermo Scientific 10X DyNAzyme buffers are available in several formats: with and without detergents as well as in Mg2+-free solution.
Includes:
Detergent-free 10x Optimized DyNAzyme EXT Buffer
Recommended for:
PCR, long PCR, DHPLC, microarray