Please login to your online account to display your discounted pricing

Thermo Scientific™ DyNAzyme I & II DNA Polymerases


Get improved thermal stability at high temperatures with these Taq-like enzymes, Thermo Scientific™ DyNAzyme™ DNA Polymerases.

Manufacturer: thermo scientific™  F501S

 View more versions of this product

Catalog No. F501S

  • / Each
 

Description & Specifications

Specifications

Includes DyNAzyme II DNA Polymerase, 10x Optimized DyNAzyme Buffer
Concentration PCR
Description DyNAzyme™ II DNA Polymerase
Storage Requirements Recommended storage temperature -20 °C. Stable for one year from the assay date.
Quantity 250U (2U/µL)

DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.

Highlights

  • Improved thermal stability
  • Improved tolerance against DMSO
  • Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
  • Detergent-free buffer also available