Please login to your online account to display your discounted pricing
Thermo Scientific™ DyNAzyme Buffers
Choose from several formats of these buffers to optimize PCR using Thermo Scientific™ DyNAzyme™ DNA Polymerases.
$19.49 - $20.46
Products
Catalog Number Mfr. No. For Use With (Application) Includes Quantity Price Quantity    
F510L
thermo scientific™
F510L
PCR Mg2+-free 10x DyNAzyme Buffer and 50mM MgCl2 Solution 4 x 1.5mL Each for $20.46
F511L
thermo scientific™
F511L
PCR 10x Optimized DyNAzyme Buffer 4 x 1.5mL Each for $20.46
F517L
thermo scientific™
F517L
PCR, long PCR, DHPLC, microarray Detergent-free 10x Optimized DyNAzyme EXT Buffer 4 x 1.5mL Each for $19.49
Description & Specifications
Optimized Thermo Scientific 10X DyNAzyme buffers are available in several formats: with and without detergents as well as in Mg2+-free solution.
Includes
Mg-free 10X DyNAzyme Buffer (F-510):
  • 1X buffer contains: 10mM Tris-HCl (pH 8.8 at 25°C), 50mM KCl, and 0.1%Triton X-100
  • 50mM MgCl2
Optimized 10X DyNAzyme Buffer (F-511):
  • 1X buffer contains 10mM Tris-HCl (pH8.8 at 25°C), 1.5mM MgCl2, 50mM KCl, and 0.1% Triton X-100
Optimized 10X DyNAzyme Buffer, Detergent-free (F-516):
  • 1X buffer contains 10mM Tris-HCl (pH8.8 at 25°C), 1.5mM MgCl2,and 50 mM KCl
Optimized 10X DyNAzyme EXT Buffer,Detergent-free(F-517):
  • 1X buffer contains 50mM Tris-HCl (pH9.0 at 25°C), 1.5mM MgCl2,and 15mM (NH4)2SO4