Please login to your online account to display your discounted pricing
Thermo Scientific™ DyNAzyme Buffers
Choose from several formats of these buffers to optimize PCR using Thermo Scientific™ DyNAzyme™ DNA Polymerases.
Manufacturer: thermo scientific™  F510L
 View more versions of this product
Catalog No. F510L
  • / Each
Description & Specifications
Specifications
Description Mg2+-free DyNAzyme™ Buffer and 50mM MgCl2 Solution
For Use With (Application) PCR
Includes Mg2+-free 10x DyNAzyme Buffer and 50mM MgCl2 Solution
Quantity 4 x 1.5mL
Optimized Thermo Scientific 10X DyNAzyme buffers are available in several formats: with and without detergents as well as in Mg2+-free solution.
Includes
Mg-free 10X DyNAzyme Buffer (F-510):
  • 1X buffer contains: 10mM Tris-HCl (pH 8.8 at 25°C), 50mM KCl, and 0.1%Triton X-100
  • 50mM MgCl2
Optimized 10X DyNAzyme Buffer (F-511):
  • 1X buffer contains 10mM Tris-HCl (pH8.8 at 25°C), 1.5mM MgCl2, 50mM KCl, and 0.1% Triton X-100
Optimized 10X DyNAzyme Buffer, Detergent-free (F-516):
  • 1X buffer contains 10mM Tris-HCl (pH8.8 at 25°C), 1.5mM MgCl2,and 50 mM KCl
Optimized 10X DyNAzyme EXT Buffer,Detergent-free(F-517):
  • 1X buffer contains 50mM Tris-HCl (pH9.0 at 25°C), 1.5mM MgCl2,and 15mM (NH4)2SO4