Please login to your online account to display your discounted pricing

Dynasore, Tocris

Non-competitive dynamin inhibitor

$164.61 - $683.95

Chemical Identifiers

Chemical Name or Material Dynasore
Assay Percent Range >99%
Molecular Formula C18H14N2O4
Formula Weight 322.32
Synonym 3-Hydroxynaphthalene-2-carboxylic acid (3, 4-dihydroxybenzylidene)hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

289710

 
r & d systems
RDS289710
10mg Each for $164.61

289750

 
r & d systems
RDS289750
50mg Each for $683.95
Description & Specifications

Specifications

Formula Weight 322.32
Quantity 10mg
Synonym 3-Hydroxynaphthalene-2-carboxylic acid (3, 4-dihydroxybenzylidene)hydrazide
Molecular Formula C18H14N2O4
Assay Percent Range >99%
CAS 304448-55-3
Chemical Name or Material Dynasore
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C