missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ DyNAmo Flash SYBR™ Green qPCR Kit

Developed for fast real-time qPCR, the kit provides qPCR results faster than most SYBR Green kits without compromising the qPCR performance.

Manufacturer:  Thermo Scientific™ F415S

 View more versions of this product

Catalog No. F415S


 

Disclaimers:

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.


Specifications

Description

A DNA-binding domain attached to the polymerase in this kit improves the physical stability of the polymerase-DNA complex, thus enhancing the processivity and robustness of the amplification. The high amplification efficiency of DyNAmo Flash SYBR Green qPCR Kit gives reliable quantification and early Cq values. Due to the high signal intensity, DyNAmo Flash SYBR Green qPCR provides an excellent signal-to-noise ratio.

  • Extremely fast protocols: Combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • Included dUTP allows the use of UNG for prevention of carry-over contamination
  • Convenient 2X master mix and optimized protocol
Description & Specifications

Specifications

Real time PCR (qPCR), Fast qPCR, qPCR using SYBR green dye, RT-qPCR using SYBR green dye
100 Reactions
Store at -20°C.
5700 Sequence Detection System, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, Bio-Rad CFX384 Touch™, BioRad CFX96™, Cepheid SmartCycler, Corbett Rotor-Gene, Eppendorf MasterCycler RealPlex, MJ Chromo4, MJ Opticon, QuantStudio™, Roche LightCycler 480, StepOne™ System, StepOnePlus™ System, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, ViiA™ 7 System
Fast
1 x 1mL DyNAmo Flash SYBR Green qPCR Mix (2X); 1 x 50μL 50X ROX dye, 25μM
40 x 50µL Rxns (100 x 20µL Rxns)
Detection Method: SYBR; PCR Method: qPCR
DNA, dsDNA