missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ DyNAmo Flash Probe qPCR Kit

Perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput.

Manufacturer:  Thermo Scientific™ F455S

 View more versions of this product

Catalog No. F455S


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: DyNAmo Flash qPCR Mix (2X); 50X ROX dye, 25μM

Specifications

Description

DyNAmo Flash Probe qPCR Kit is the perfect choice for fast probe-based qPCR. The optimized reagents and extremely short cycling times deliver maximum speed thus increasing sample throughput. Significant time savings are achieved with both conventional and fast real-time PCR instruments. Performance of the DyNAmo Flash Probe qPCR Kit is based on an efficient hot start Thermus brockianus DNA polymerase. Sensitivity and the excellent amplification efficiency across a wide range of template concentrations guarantee reproducible results.

DyNAmo Flash Probe qPCR Kit is optimized for hydrolysis probes (e.g., TaqMan probes) and it can be used with both block- and capillary-based realtime PCR instruments.

  • Extremely fast protocols: combined annealing and extension step of only 15 sec.
  • Specific and sensitive detection of a wide range of template concentrations
  • Included dUTP allows the use of UNG for prevention of carry-over contamination
  • Convenient 2X master mix and optimized protocols
Description & Specifications

Specifications

DyNAmo™ Flash Probe qPCR Kit
1 x 1mL DyNAmo Flash qPCR Mix (2X); 1 x 50μL 50X ROX dye, 25μM
40 x 50µL Rxns (100 x 20µL Rxns)
Primer-Probe Detection
DNA, dsDNA
Real time PCR (qPCR), Fast qPCR, qPCR using sequence-specific probes, RT-qPCR using sequence-specific probes
100 Reactions
Store at -20°C.
5700 Sequence Detection System, 7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, Bio-Rad CFX384 Touch™, BioRad CFX96™, BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5, Cepheid SmartCycler, Corbett Rotor-Gene, Eppendorf™ MasterCycler™ RealPlex, MJ Chromo4, MJ Opticon, QuantStudio™, Roche LightCycler 480, StepOne™ System, StepOnePlus™ System, Stratagene Mx3000P, Stratagene Mx3005P, Stratagene Mx4000, ViiA™ 7 System
Fast