Dynalon™ Heavy Walled HDPE Carboys
$72.06 - $422.55
Specifications
Autoclavable No
Handles Swing
Material HDPE
For Use With (Application) Heavy-duty swing handle allows easy carrying of liquids and powders
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Inner Neck Diameter (Metric) Outer Capacity (English) Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Neck Diameter (Metric) Outer Capacity (English) Capacity (Metric) Price Quantity  
14-400-101 Dynalon™
2086050010
5.5cm 21.1cm 338.14 oz. 10L Each for $136.00
14-400-103 Dynalon™
2086050050
5.5cm 35.07cm 1690.7 oz. 50L Each for $422.55
14-400-107 Dynalon™
2086850005
4.6cm 16.3cm 169.07 oz. 5L Each for $72.06
14-400-108 Dynalon™
2086850010
5.5cm 21cm 338.14 oz. 10L Each for $110.67
14-400-109 Dynalon™
2086850025
5.5cm 37.9cm 845.35 oz. 25L Each for $171.82
14-400-110 Dynalon™
2086850050
5.5cm 36.6cm 1690.7 oz. 50L Each for $359.85
14-400-100 Dynalon™
2086050005
4.6cm 16.02cm 169.07 oz. 5L Each for $103.27
14-400-102 Dynalon™
2086050025
5.5cm 27.94cm 845.35 oz. 25L Each for $218.42
Specifications Description & Specifications
Specifications
No
Swing
HDPE
Heavy-duty swing handle allows easy carrying of liquids and powders