Dynalon™ Bio-Bin™ Waste Disposal Containers
A new cost effective and convenient alternative to rigid plastic waste collection and disposal containers
Manufacturer:  Dynalon™ 7973030030
 View more versions of this product
Catalog No. 12-009-802
Description
Description
  • Space efficient, light weight fold-flat design
  • Built-in lid and handles
  • Waterproof construction
  • Temporary and permanent seals
  • Built-in super absorbent Bio-matt™ to soak up any excess liquid
  • Autoclavable (use with #202065 Autoclave Sterility Tape)
  • Supplied with 2 x 2 in. Biohazard Waste Labels
  • Stackable post use (UN 3291 Approved)
Order Info
Accessories available. (See Cat. No. 12-009-803)
Specifications
Specifications
30 L
7.9 gal.
Waste Container
27.94 cm
2 x 2 in. biohazard waste labels
11 in.
21 in.
UN 3291
11 x 11 x 21 in. (27.94 x 27.94 x 53.34 cm)
53.34 cm
27.94 cm
Ideal solution for biohazard or non-hazardous waste
11 in.