Please login to your online account to display your discounted pricing

Thermo Scientific™ DyLight™ Fluor Phosphine Reagents


Label and detect azide-tagged molecules for metabolic labeling (Staudinger ligation) for fluorescence assays with phosphine-activated fluorescent dyes.

Manufacturer: thermo scientific™  PI88907

 View more versions of this product

Catalog No. PI88907

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Formulation Phosphine-activated proprietary green fluorescent dye
Sufficient For 1 use to label a total of 6mg of IgG using typical conditions
Description DyLight 488-Phosphine

Thermo Scientific™ DyLight Fluor Phosphine Reagents are phosphine-activated fluorescent dyes for specific labeling and detection of azide-tagged molecules, which enables use of fluorescence imaging in metabolic labeling strategies.

Azide groups, which are the reactive targets of the phosphine-activated fluors, can be introduced into proteins or other cellular targets via azide reagents or through in vivo labeling with azide-derivatives of naturally occurring metabolic building blocks. Because neither phosphines nor azides are present in biological systems, they comprise chemoselective (mutually specific) ligation pairs for labeling and conjugation. Thus, when used in combination with azide labeling strategies, Phosphine-Activated DyLight Fluors enable directed fluorescent labeling to facilitate detection of protein interactions and post-translational modifications using fluorescence imaging technologies.

Highlights:

  • Soluble easily dissolves in water-miscible solvents (e.g., DMSO) for subsequent dilution in aqueous reaction mixtures with cell lysates and other biological samples

  • Compatible reaction chemistry occurs effectively in simple buffer conditions; requires no accessory reagents such as copper or reducing agents, and does not interfere with fluorescence applications

  • Chemoselective the phosphine reactive group is specific in biological samples for azide-tagged molecules, ensuring that fluorescent labeling is specific

  • High-performance fluorescence DyLight 488, DyLight 550 and DyLight 650 are intense, highly stable fluorophores for green, orange and red fluorescent detection