Please login to your online account to display your discounted pricing

Thermo Scientific™ DyLight™ Fluor Antibody Labeling Kits


Label and purify antibodies for fluorescence imaging assays in one hour with the full spectral range of activated DyLight Fluorescent Dye kits.

Manufacturer: thermo scientific™  62276

 View more versions of this product

Catalog No. PI62276

  • / Each
 

Description & Specifications

Specifications

Formulation Multi-component kit with proprietary blue fluorescent dye
Sufficient For Labeling and purifying 5 x 100ug (1mg/mL) of IgG or other proteins
Kit Contents Borate Buffer (0.67M), 1mL
Quantity 5 rxn kit
Description DyLight 350 Antibody Labeling Microscale Kit

Thermo Scientific™ DyLight Fluor Antibody Labeling Kits provide the essential reagents and accessories to easily label and purify antibodies and include regular-scale and microscale kits for each specific DyLight Fluorescent Dye.

Two convenient kit formats are available to accommodate varied labeling requirements. The DyLight Antibody Labeling Kits contain all necessary components to perform three separate labeling reactions using 1mg of IgG or similar quantities of other proteins. The DyLight Microscale Antibody Labeling Kits contain all the necessary components to perform five separate labeling reactions using 100μg of IgG. The labeling kits use high-performance spin desalting columns to provide exceptional dye removal and antibody recovery.

Highlights:

  • Fast - label and purify protein in approximately one hour
  • Amine-reactive dyes - label virtually any protein
  • Efficient non-reacted dye removal
  • Minimal sample dilution
  • Spin column format eliminates the need for column preparation, fraction screening and waiting for protein to emerge from column

Item-Specific Features:

  • Standard kits contain sufficient reagents to label and purify 3 × 1mg of IgG or similar quantities of other proteins
  • Microscale kits contain sufficient reagents to label and purify 5 × 0.1mg of IgG or similar quantities of other proteins

Includes:

Antibody Labeling Kits

DyLight NHS Ester, 3 vials; Reaction buffer, 1mL; Spin columns, 6 each; Microcentrifuge collection tubes, 12 each; Purification resin, 5mL

Microscale Kits

DyLight NHS Ester, 5 vials; Reaction buffer, 1mL; Spin columns, 5 each; Microcentrifuge collection tubes, 10 each; Purification resin, 5mL