Please login to your online account to display your discounted pricing

Thermo Scientific™ DyLight™ 680-4xPEG Amine-Reactive Dye and Kits


Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.

Manufacturer: thermo scientific™  53077

 View more versions of this product

Catalog No. PI53077

  • / Each
 

Description & Specifications

Specifications

Description DyLight 680-4xPEG Microscale Antibody Labeling Kit
Sufficient For Labeling and purifying 5 x 100µg (1mg/mL) of IgG or other proteins
Quantity 5-rxn kit
Kit Contents Borate Buffer (0.67M), 1mL
Formulation Multi-component kit with proprietary near-infrared fluorescent dye

Fluorescently label antibodies and other proteins with Thermo Scientific™ DyLight 680-4xPEG Amine-Reactive Dye, a derivative of our high-performance DyLight 680 Dye.


Additional Information and Online Ordering »

The DyLight 680-4xPEG dye contains 4 polyethylene glycol (PEG) chains that are non–cytotoxic, enhance fluorescence and reduce nonspecific binding of conjugates made with them. Conjugates made with DyLight 680-4xPEG Dye can be used as molecular probes for cellular imaging and other fluorescence detection methods. The far-red to NIR fluorescence properties of DyLight 680-4xPEG Dye make it especially useful in a variety of biological, chemical, and pharmaceutical applications, including in vivo imaging. The PEG chains also improve solubility of the dyes and labeled molecules in aqueous solution, aid in cell permeability and improve tissue retention.

Highlights:

  • High fluorescence intensity – significantly brighter fluorescence than Alexa Fluor™ 680

  • PEGylated – improves solubility in aqueous solution, aids in cell permeability and improves tissue retention in in vivo applications

  • Convenient kit sizes – standard and microscale kit sizes to match your experimental needs

Recommended for:

  • Fluorescence microscopy
  • In vivo or ex vivo imaging
  • Cell-based assays
  • Flow cytometry/fluorescence-activated cell sorting (FACS)