missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dyclonine hydrochloride

$379.36 - $379.36

Chemical Identifiers

CAS 536-43-6
Molecular Formula C18H28ClNO2
Molecular Weight (g/mol) 325.877
MDL Number MFCD00035386
InChI Key KNZADIMHVBBPOA-UHFFFAOYSA-N
Synonym dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, dyclonine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride
PubChem CID 68304
ChEBI CHEBI:4725
IUPAC Name 1-(4-butoxyphenyl)-3-piperidin-1-ylpropan-1-one;hydrochloride
SMILES CCCCOC1=CC=C(C=C1)C(=O)CCN2CCCCC2.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6133014 View Documents Alfa Aesar™
J6133014
25g Each for $379.36
 
Description

Description

Applications
Dyclonine hydrochloride is used for the temporary relief of the following occasional mouth and throat symptoms: pain, minor irritation, sore mouth and sore throat. It is a sodium channel blocker and a HSP90 chaperone protein inhibitor. It shows local anesthetic effects in vivo and is orally active.

Solubility
Partly soluble in water. Soluble in chloroform, ethanol, acetone and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

536-43-6
325.877
KNZADIMHVBBPOA-UHFFFAOYSA-N
68304
1-(4-butoxyphenyl)-3-piperidin-1-ylpropan-1-one;hydrochloride
C18H28ClNO2
MFCD00035386
dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, dyclonine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride
CHEBI:4725
CCCCOC1=CC=C(C=C1)C(=O)CCN2CCCCC2.Cl

Specifications

White
25g
C18H28ClNO2
14,3473
KNZADIMHVBBPOA-UHFFFAOYSA-N
1-(4-butoxyphenyl)-3-piperidin-1-ylpropan-1-one;hydrochloride
68304
325.88
Dyclonine hydrochloride
173°C to 178°C
536-43-6
MFCD00035386
dyclonine hydrochloride, dyclonine hcl, dyclone, dyclothane, tanaclone, 4'-butoxy-3-piperidinopropiophenone hydrochloride, dyclocaine hydrochloride, dyclonine hydrochloride, unii-zec193879q, 1-propanone, 1-4-butoxyphenyl-3-1-piperidinyl-, hydrochloride
CCCCOC1=CC=C(C=C1)C(=O)CCN2CCCCC2.Cl
325.877
CHEBI:4725
Crystalline Powder
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : UG8750000