Please login to your online account to display your discounted pricing

DV 7028 hydrochloride, Tocris™

Selective 5-HT2A antagonist; antithrombotic

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material DV 7028 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H25FN4O3·HCl
Formula Weight 436.91
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2H-pyrido[1,2-a]-1,3,5-triazine-2,4(3H)-dione hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

448050

 
r & d systems
RDS448050
50mg Each for $794.96

448010

 
r & d systems
RDS448010
10mg Each for $188.60
Description & Specifications

Specifications

Formula Weight 436.91
Quantity 50mg
Chemical Name or Material DV 7028 hydrochloride
Recommended Storage Desiccate at Room Temperature
CAS 133364-62-2
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2H-pyrido[1,2-a]-1,3,5-triazine-2,4(3H)-dione hydrochloride
Molecular Formula C21H25FN4O3·HCl
Assay Percent Range >99%
Safety and Handling

Desiccate at Room Temperature