Please login to your online account to display your discounted pricing

Duroquinone 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  117710050

 View more versions of this product

Catalog No. AC117710050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥96.0%
Formula Weight 164.2
Quantity 5g
Color Yellow
RTECS Number GU5410000
EINECS Number 208-409-8
Melting Point 109° to 114°C
Beilstein 07, 669
Merck Index 15, 3517
Infrared Spectrum Authentic
Chemical Name or Material Duroquinone
Name Note 97%
Assay Percent Range 97%
Molecular Formula C10H12O2
Packaging Glass bottle
MDL Number MFCD00001604
CAS 527-17-3
Synonym Tetramethyl-p-benzoquinone

Specifications

Chemical Name or Material Duroquinone
Name Note 97%
CAS 527-17-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H12O2
Formula Weight 164.2
MDL Number MFCD00001604
Synonym Tetramethyl-p-benzoquinone