Please login to your online account to display your discounted pricing

Duroquinone 97%, ACROS Organics™

$99.99 - $99.99

Chemical Identifiers

Chemical Name or Material Duroquinone
Name Note 97%
CAS 527-17-3
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C10H12O2
Formula Weight 164.20g/mol
MDL Number MFCD00001604
Synonym Tetramethyl-p-benzoquinone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC117710050

  SDS
fisher scientific acros
117710050
Glass bottle 5g Each for $99.99
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Melting Point 109° to 114°C
Quantity 5g
Chemical Name or Material Duroquinone
Name Note 97%
CAS 527-17-3
Physical Form Yellow powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C10H12O2
Formula Weight 164.20g/mol
MDL Number MFCD00001604
Beilstein 07, 669
Merck Index 15, 3517
Synonym Tetramethyl-p-benzoquinone
RTECS Number GU5410000
EINECS Number 208-409-8