DWK Life Sciences DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring
$313.94 - $1,299.26
Specifications
Color Clear
Material Glass
View More Specs
Products 4
Catalog Number Mfr. No. Capacity Diameter (Metric) Height (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity Diameter (Metric) Height (Metric) Price Quantity  
09-841-176 View Documents DWK Life Sciences
218013651
250 mL 70mm 143mm Case for $313.94
09-841-177 View Documents DWK Life Sciences
218014459
500 mL 86mm 181mm Case for $392.63
09-841-179 View Documents DWK Life Sciences
218015455
1000 mL 101mm 230mm Case of 10 Each for $559.46
09-841-180 View Documents DWK Life Sciences
218016357
2000 mL 136mm 265mm Case for $1,299.26
Description
Description
  • Service temperature level of screw cap and pouring ring: +140 °C
  • Typical applications: storage, sample preparation, transport
  • Select sizes available with retrace code
Specifications
Specifications
Clear
Glass