Please login to your online account to display your discounted pricing
H-B Instrument™ Durac™ Specific Gravity Precision Hydrometers
Used in food and beverage, chemical, pharmacology, transportation, refrigeration industries
Manufacturer: h-b instrument™  B618011200
 View more versions of this product
Catalog No. 13-202-801
  • / Each
  • / Case of 10 Each
Description & Specifications
Specifications
Length (Metric) 325mm
Accuracy 0.001 sp. gr.
Specific Gravity Range 1.300/1.370
Temperature of Standardization 15°C (60°F)
Ballast Type Lead free
Quantity 10/Pk.
Product Type Specific Gravity Precision Hydrometer