H-B Instrument™ Durac™ Plus™ Individually Calibrated ASTM Liquid-In-Glass Thermometers, Organic Liquid Fill
Use these mercury-free, calibrated thermometers in quality control laboratories. The Durac™ Plus™ ASTM Liquid-In-Glass Thermometer conforms to ASTM standards for methods specified in the order table.
Manufacturer:  H-B Instrument™ HBB607800600
 View more versions of this product
Catalog No. 13-203-249
Description
Description
  • Individually calibrated at specified points and supplied with a Calibration Report stating corrections to one-tenth of the smallest division
  • Ideal for use in quality control laboratories
  • Blue organic liquid fill against lead-free white back glass; individually serialized
Warranty
One year limited
Specifications
Specifications
S65C-03 / Precision
50°C to 80°C
50°C
Total
Blue Organic Liquid
±0.1°C
0.1°C
80°C
0, 50, 60, 70, 80°C
405 mm
ASTM Thermometer
Documents