DuPont™ Tyvek™ Lab Coats
Manufacturer:  DuPont™ TY210SWHXL003000
Catalog No. 01-361-11D
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Description
Disposable, yet strong enough to withstand several wearings
Provide excellent protection against submicron particles.
  • Lightweight and tear-resistant Tyvek fabric
  • Knee length with five-snap front and full sleeves
  • Chemically and biologically inert
  • Provide limited splash protection
  • White
Applications:
Lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Specifications
Specifications
Lay down
White
Tyvek™ Fabric
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
5.08 to 6.02 ft.
34.75 in.
Serged
Knee length
Lab Coat with No Pockets, Open Wrists
Snap
Open
X-Large
Yes
1.54 to 1.83m
88.26cm
41.75 to 45.25 in.
Bulk