missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tyvek™ IsoClean™ Series 253 Coveralls, Sterile
Excellent for a controlled environment to protect against particulates and other contaminants.
Manufacturer: DuPont™ IC253BWHMD00250S
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
- Garments are gamma sterilized to SAL of 10-6
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Coverall has elastic openings for tighter fit at wrist and ankle
- Bound neck for lower particle shedding
- Front zipper closure for easy donning and doffing
- Dolman sleeve design for greater range of motion and comfort
- Full traceability on all sterilized apparel with certificates of sterility available
Specifications
Covered,Elastic | |
Tyvek™ IsoClean™ Flashspun Polyethylene | |
Bound | |
Covered,Elastic | |
5.5 to 5.10 ft. | |
35.5 in. | |
Collared | |
Medium | |
30.5 in. | |
77.4 cm | |
5 mil |
Zipper | |
Sterile | |
White | |
Excellent barrier to dry particles, microorganisms and non-hazardous liquids | |
167.64 to 155.44 cm | |
90.17 cm | |
Bound | |
36 to 38 in. | |
91.44 to 96.52 cm | |
Dolman |