DuPont™ Tyvek™ 127S Series Coveralls
Manufacturer:  DuPont™ TY127SWH3X002500
 View more versions of this product
Catalog No. 01-361-7F
Description
Description
Tyvek 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. The fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away.
  • Comfort fit design based on extensive wearer input to provide our most comfortable garment design that: enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes a longer zipper for easier donning and doffing and an elastic waist to better position the garment.
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Longer zipper extends to chin for complete coverage of neck area
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
Compliance
Meets USDA, FDA, OSHA standards; ASTM™ D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.
Specifications
Specifications
USDA, FDA, OSHA, ASTM D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127
Elastic
White
6.2 to 6.4 ft.
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
97.79cm
81.28cm
Zipper front
Hooded
HDPE
49.25 to 52.75 in.
Lay down
Serged
3X-Large
1.88 to 1.95m
38.5 in.
32 in.
5.7mil
Elastic
Yes
125 to 133.98cm
Documents