missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tyvek™ 125S Series Coveralls

Coveralls with elastic wrists and ankles. Tyvek 400 provides an ideal balance of protection, durability and comfort. Design enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts.

Manufacturer:  DuPont™ TY125SWHLG002500

 View more versions of this product

Catalog No. 01-361-4C



Description

Description

  • Fabric: Tyvek 400D; serged seams
  • Tyvek 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material that provides durability and comfort
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Laydown collar; longer zipper extends to chin for complete coverage of neck area
  • Storm flap of garment material covers zipper
  • Elastic opening for tighter fit at wrists and ankles
  • Elastic waist to better position the garment
  • Thumb loop to keep garment sleeve in place
  • Applications include fiberglass and composite manufacturing, wind turbines, boat construction and repair, remediation, and maintenance

Compliance

Meets ASTM™ D1777 standards; AATCC 127 compliant.

Specifications

Specifications

Elastic
HDPE
Lay down
Elastic
Yes
5.5 to 5.9 ft.
35.5 in.
Collared
Large
29.5 in.
74.93 cm
Front zipper
ASTM D1777, AATCC 127
White
Flashspun HDPE
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
1.67 to 1.8 m
90.17 cm
Serged
39.75 to 43.25 in.
100.96 to 109.85 cm
5.7 mil
SDS
Documents
Product Certifications
Promotions

Promotions