DuPont™ Tyvek™ 122S Series Coveralls
Comfort fit design based on extensive wearer input to provide a comfortable garment design that enables a greater range of movement while stretching and bending.
Manufacturer:  DuPont™ TY122SWH4X002500
 View more versions of this product
Catalog No. 01-361-2G
Alert:
Garments made of Tyvek should not be used around heat, flames, sparks, or in potentially flammable or explosive environments.
Description
Description
Comfort fit design based on extensive wearer input to provide a comfortable garment design that enables a greater range of movement while stretching and bending.
  • Comfort fit design provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Attached Tyvek FC boots have special coating to provide added skid-resistance
  • Longer zipper extends to chin for complete coverage of neck area and provides easeir donning and doffing
  • Elastic opening for tighter fit at wrist
  • Elastic waist to better position garment
  • Elastic ankles
  • Serged seam
  • White
Compliance
Meets USDA, FDA, OSHA standards; ASTM™ D1777, ASTM D3776, ASTM D774, ASTM D5733, ASTM D5034, ASTM D257, AATCC 127 compliant.
Specifications
Specifications
Elastic
White
6.4 to 6.7 ft.
Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
97.8cm
82.5cm
33 in.
83.8cm
Attached boots,Elastic
Attached skid-resistant boot
133.9 to 142.8cm
Serged
4X-Large
1.9 to 2.0m
38.5 in.
32.5 in.
5.9 mils
Elastic
Zipper front
Yes
Flash spun high-density polyethylene
52.75 to 56.25 in.
Documents