Please login to your online account to display your discounted pricing

DuPont™ Tyvek™ Bib Apron and Sleeves

Protection against many hazardous powders

$117.31 - $188.81

Specifications

Color White
Material Tyvek™ Fabric
Size Universal
For Use With (Application) Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up
View More Specs

 Alert

Garments made of Tyvek should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Products
Catalog Number Mfr. No. Seam Type Length (English) Length (Metric) Price Quantity    

01-361-21

 
dupont canada inc
TY500SWH00020000
Serged 18 in. 45.7cm Case of 100 Pair for $117.31

01-361-83

 
dupont canada inc
TY273BWH00010000
Bound 36 in. 69cm Case of 100 Each for $188.81
Description & Specifications

Specifications

Color White
Material Tyvek™ Fabric
Size Universal
For Use With (Application) Lead and asbestos abatement/remedation, general maintenance/operations, spray painting, general clean-up

Nonwoven Tyvek fabric helps resist penetration by particles and other contaminants. The inherent barrier stops submicron particles even after abrasion.

Nonwoven Tyvek fabric helps resist penetration by particles and other contaminants. The inherent barrier stops submicron particles even after abrasion.

Apron

  • Provides coverage of front and sides
  • With bound neck and ties
  • White
Sleeves
  • Offer wrist-to-elbow protection
  • Elasticized seams at both ends
  • White