DuPont™ Tychem™ ThermoPro™ TP198T Coveralls
ThermoPro coveralls offer superior protection against fire, electricity, and toxic chemicals.
Manufacturer:  DuPont™ TP198TGY3X0002BN
 View more versions of this product
Catalog No. 19-101-303
Description
Description
ThermoPro coveralls are secure in fit to seal out contaminants
  • Respirator fit hood with drawstrings
  • Front zipper with storm flap, hook and loop secure
  • Elastic wrists
  • Open, hemmed ankles
  • Provides at least one hour breakthrough protection against 20 of the 21 standard ASTM F1001 chemicals
  • At least 30 minutes of protection against 182 chemical challenges
  • Protects against flash fire, electric arc and liquid-chemical splash, wide range of toxic and chemical warfare agents
  • HazMat teams in fire departments
  • Industrial fire brigades
  • Clandestine lab investigation
  • Industrial chemical processing plants
Certifications
  • NFPA 1992 (2012)
  • NFPA 2112 (2012)
  • Exceeds NFPA 70E Hazard Risk Category 2
Specifications
Specifications
Open
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
Gray
Fit hood, elastic wrist, open ankle, with respirator; HDPE Tychem™ ThermoPro construction
6.2 to 6.4 ft.
39 in.
Hooded
3X-Large
29.75 in.
31.75 in.
HDPE
Hook and loop
Elastic
Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants
1.88 to 1.95 m
99 cm
Taped
50.25 to 53.75 in.
127.63 to 136.52 cm