DuPont™ Tychem™ ThermoPro™ TP198T Coveralls
ThermoPro coveralls offer superior protection against fire, electricity, and toxic chemicals.
Manufacturer:  DuPont™ TP198TGYLG000200
 View more versions of this product
Catalog No. 19-101-307
Description
Description
ThermoPro coveralls are secure in fit to seal out contaminants
  • Respirator fit hood with drawstrings
  • Front zipper with storm flap, hook and loop secure
  • Elastic wrists
  • Open, hemmed ankles
  • Provides at least one hour breakthrough protection against 20 of the 21 standard ASTM F1001 chemicals
  • At least 30 minutes of protection against 182 chemical challenges
  • Protects against flash fire, electric arc and liquid-chemical splash, wide range of toxic and chemical warfare agents
  • HazMat teams in fire departments
  • Industrial fire brigades
  • Clandestine lab investigation
  • Industrial chemical processing plants
Certifications
  • NFPA 1992 (2012)
  • NFPA 2112 (2012)
  • Exceeds NFPA 70E Hazard Risk Category 2
Specifications
Specifications
Open
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™
Gray
Fit hood, elastic wrist, open ankle, with respirator; HDPE Tychem™ ThermoPro construction
5.5 to 5.9 ft.
35 in.
Hooded
Large
27.5 in.
26 in.
HDPE
Hook and loop
Elastic
Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants
1.67 to 1.8 m
88.9 cm
Taped
38.75 to 42.25 in.
98.42 to 107.31 cm