missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ ThermoPro™ TP198T Coveralls
ThermoPro coveralls offer superior protection against fire, electricity, and toxic chemicals.
Manufacturer: DuPont™ TP198TORXL000200
Description
ThermoPro coveralls are secure in fit to seal out contaminants
- Respirator fit hood with drawstrings
- Front zipper with storm flap, hook and loop secure
- Elastic wrists
- Open, hemmed ankles
- Provides at least one hour breakthrough protection against 20 of the 21 standard ASTM F1001 chemicals
- At least 30 minutes of protection against 182 chemical challenges
- Protects against flash fire, electric arc and liquid-chemical splash, wide range of toxic and chemical warfare agents
- HazMat teams in fire departments
- Industrial fire brigades
- Clandestine lab investigation
- Industrial chemical processing plants
Certifications
- NFPA 1992 (2012)
- NFPA 2112 (2012)
- Exceeds NFPA 70E Hazard Risk Category 2
Specifications
Open | |
HDPE | |
Orange | |
Fit hood, elastic wrist, open ankle, with respirator; HDPE Tychem™ ThermoPro construction | |
5.8 to 6.2 ft. | |
36.5 in. | |
Hooded | |
X-Large | |
28.25 in. | |
28 in. |
Hook and loop | |
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™ | |
Elastic | |
Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants | |
1.76 to 1.88 m | |
92.7 cm | |
Taped | |
42.75 to 46.25 in. | |
108.58 to 117.47 cm |