DuPont™ Tychem™ ThermoPro™ TP198T Coveralls
ThermoPro coveralls offer superior protection against fire, electricity, and toxic chemicals.
Manufacturer:  DuPont™ TP198TORMD0002BN
 View more versions of this product
Catalog No. 19-101-325
Description
Description
ThermoPro coveralls are secure in fit to seal out contaminants
  • Respirator fit hood with drawstrings
  • Front zipper with storm flap, hook and loop secure
  • Elastic wrists
  • Open, hemmed ankles
  • Provides at least one hour breakthrough protection against 20 of the 21 standard ASTM F1001 chemicals
  • At least 30 minutes of protection against 182 chemical challenges
  • Protects against flash fire, electric arc and liquid-chemical splash, wide range of toxic and chemical warfare agents
  • HazMat teams in fire departments
  • Industrial fire brigades
  • Clandestine lab investigation
  • Industrial chemical processing plants
Certifications
  • NFPA 1992 (2012)
  • NFPA 2112 (2012)
  • Exceeds NFPA 70E Hazard Risk Category 2
Specifications
Specifications
Open
HDPE
Orange
Fit hood, elastic wrist, open ankle, with respirator; HDPE Tychem™ ThermoPro construction
5.3 to 5.7 ft.
34 in.
Hooded
Medium
26.5 in.
24 in.
Hook and loop
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
Elastic
Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants
1.6 to 1.73 m
86.3 cm
Taped
34.75 to 38.25 in.
88.26 to 97.15 cm