DuPont™ Tychem™ ThermoPro™ Sleeveless Aprons
Designed to provide protection for industrial workers and Hazmat responders. DuPont™ Tychem™ ThermoPro™ Sleeveless Aprons provide permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents, and provide escape time from flash fires and protection from electric arc.
$686.57 - $731.72
Specifications
For Use With (Application) HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories
Seam Type Taped
Sleeve Style Sleeveless
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Size Price Quantity  
Catalog Number Mfr. No. Color Size Price Quantity  
19-122-000 DuPont™
TP284TGYSM000400
Gray Small Case of 4 Each for $731.72
19-122-001 DuPont™
TP284TGYMD000400
Gray Medium Case of 4 Each for $731.72
19-122-002 DuPont™
TP284TGYLG000400
Gray Large Case of 4 Each for $731.72
19-122-006 DuPont™
TP284TORLG000400
Orange Large Case of 4 Each for $731.72
19-122-005 DuPont™
TP284TORMD000400
Orange Medium Case of 4 Each for $731.72
19-122-003 DuPont™
TP284TGYXL000400
Gray X-Large Case of 4 Each for $731.72
19-122-004 DuPont™
TP284TORSM000400
Orange Small Case of 4 Each for $731.72
19-122-007 DuPont™
TP284TORXL000400
Orange X-Large Case of 4 Each for $731.72
19-141-761 DuPont™
TP284TOR2X000400
Orange 2X-Large Case of 4 Each for $686.57
Specifications
Description
  • Tychem™ ThermoPro for triple hazard protection
  • Sleeveless apron design provides frontal body protection
  • For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
  • Sewn with Nomex™ thread
  • Designed with Nomex waist strap and flame resistant (FR) nylon buckle
  • Designed with end-user input for laboratory applications
  • Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
  • Gray color for discretionary needs
  • Made in the USA
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).
Description & Specifications
Specifications
HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories
Taped
Sleeveless