DuPont™ Tychem™ ThermoPro™ Sleeveless Gray Aprons
Designed to provide protection for industrial workers and Hazmat responders. DuPont™ Tychem™ ThermoPro™ Sleeveless Aprons provide permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents
Manufacturer: DuPont™ TP284TGYLG000400
Description
- Tychem™ ThermoPro for triple hazard protection
- Sleeveless apron design provides frontal body protection
- For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
- Sewn with Nomex™ thread
- Designed with Nomex waist strap and flame resistant (FR) nylon buckle
- Designed with end-user input for laboratory applications
- Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
- Gray color for discretionary needs
- Made in the USA
Order Info
Ansell™ Vinyl Closed-Back Aprons, Waist ties
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).
Specifications
HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories | |
Taped | |
Gray | |
Large |