missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont™ Tychem™ ThermoPro™ Coveralls

Provides chemical, flash fire and electric arc protection

Manufacturer:  DuPont™ TP199TGY5X0002BN

 View more versions of this product

Catalog No. 19-101-336


Alert:

Do not use for protection against continuous thermal loads, molten metal splash, hot liquids or steam. Do not wear for structural or proximity fire fighting.


Description

Description

Coveralls combine the chemical protection of DuPont Tychem with the flame and arc flash protection of DuPont Nomex™ into a single garment. The fabric delivers permeation protection against a broad range of toxic industrial chemicals and even chemical warfare agents.

  • Provide at least eights hours barrier to 20 of the 21 chemicals found in ASTM™ F1001
  • Attached respirator-fit hood with drawstring
  • Zipper front with Velcro™ fastener double storm flaps fastened by flame-resistant treated polyamide hook-and-loop material
  • Elastic at the wrists and ankles
  • Attached socks and boot flaps
  • Available in high-contrast orange or low-visibility gray
Specifications
  • Arc rating:15.2 cal/cm2
  • Thickness: 34mil
  • Basis Weight: 841 oz./yd.2
  • Ball Burst: 167 lbf
  • Tear Resistance–Trap Tear (MD/CD): 25/32 lbf
  • Breaking Strength–Grab (MD/CD): 155/170 lbf
  • Wearing Apparel Flammability: Class 1

† For electric arc hazard exposures, wear the proper arc rated garment system as dictated by an electric arc hazard analysis.

Specifications

Specifications

Attached socks,Boot overflaps
Tychem™ ThermoPro
NFPA 1992, NFPA 2112, HRC 2, NFPA 70E™, Berry Amendment
Elastic
Fire departments, industrial fire brigades, clandestine lab investigation, industrial chemical processing plants
2.04 to 2.08 m
106.6 cm
Taped
59.75 to 63.25 in.
151.76 to 160.65 cm
0.51 lb.
0.23 kg
Zipper with hook-and-loop seal and double storm flap
15.2 Calories/cm2
Gray
Respirator fit, drawstring hood, elastic wrist, attached socks with outer boot flaps, double storm flap with hook and loop closure
6.7 to 6.83 ft.
42 in.
Hooded
5X-Large
33 in.
34 mil
36.5 in.
SDS
Documents