DuPont™ Tychem™ ThermoPro Combo Suits
Protection from chemicals, flash fire and electrical arc
Manufacturer: DuPont™ TP750TORSM000200
Includes:
- Jacket with taped seams, mandarin collar, elastic wrists, front zipper closure and double storm flap
- Bib Overall with taped seams and adjustable webbing straps with closure
Description
- Constructed for heavy use, yet lightweight and easy to wear
- At least 30 minutes of protection against more than 180 chemical challenges
- Arc rating of 15.22 cal/cm Ebt
Specifications
Open | |
Mandarin | |
Small |
Elastic | |
Orange |