Learn More
DuPont™ Tychem™ SL 122 Series Coveralls, Berry Amendment Compliant
Delivers effective protection against a range of chemical environments.
$423.30 - $554.78
Specifications
Material | Tyvek&trade with Saranex&trade 23-P film laminate |
---|---|
Description | Attached hood, elastic wrist, attached socks |
Cuff Style | Elastic |
Closure Type | Front zipper |
Color | White |
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog Number | Mfr. No. | Size | Seam Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Seam Type | Price | Quantity | ||||
19-162-769
|
DuPont™
SL122TWH3X0006BN |
3X-Large | Taped |
Case of 6 Each for $475.64
|
|
19-162-768
|
DuPont™
SL122TWH2X0006BN |
2X-Large | Taped |
Case of 6 Each for $448.73
|
|
19-162-770
|
DuPont™
SL122TWH4X0006BN |
4X-Large | Taped |
Case of 6 Each for $513.72
|
|
19-162-772
|
DuPont™
SL122TWH5X0006BN |
5X-Large | Taped |
Case of 6 Each for $554.78
|
|
19-162-775
|
DuPont™
SL122TWHMD0006BN |
Medium | Taped |
Case of 6 Each for $423.30
|
|
19-162-774
|
DuPont™
SL122TWHLG0006BN |
Large | Taped |
Case of 6 Each for $423.30
|
|
19-162-776
|
DuPont™
SL122TWHXL0006BN |
X-Large | Taped |
Case of 6 Each for $423.30
|
|
Description
Tychem SL, utilizes Saranex™ 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion.
The fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Attached hood with elastic around face opening
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
Specifications
Tyvek&trade with Saranex&trade 23-P film laminate | |
Elastic | |
White | |
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments |
Attached hood, elastic wrist, attached socks | |
Front zipper | |
Non-sterile |