missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ SL 122 Series Coveralls, Berry Amendment Compliant
Delivers effective protection against a range of chemical environments.
Supplier: DuPont™ SL122TWH4X0006BN
Alert:
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Description
Tychem SL, utilizes Saranex™ 23-P film laminated to Tyvek fabric to provide a rugged and durable fabric that resists abrasion.
The fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Attached hood with elastic around face opening
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
Specifications
Hooded Coveralls with Socks | |
Tyvek with Saranex 23-P Film Laminate | |
Single-use | |
Elastic | |
Attached, Elastic Face | |
Taped | |
Attached hood, elastic wrist, attached socks | |
32 in. | |
12 mil | |
39.5 in. | |
32 in. | |
6.33 to 6.58 ft. |
4X-Large | |
White | |
Attached Socks | |
Hooded | |
Front Zipper | |
Non-sterile | |
50.75 to 54.25 in. | |
81.28 cm | |
Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments | |
100.33 cm | |
81.28 cm | |
1.92 to 2.01 m |