DuPont ™ Tychem ™ CPF 3 Coveralls with Respirator Fit Hood
Tychem CPF3 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals
Manufacturer:  Eidca Specialty Products C3128TTN2X00062K
Catalog No. 19-715-266
Description
Description
Tychem CPF3 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate
T
  • aped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Attached hood with elastic around face opening
  • Strong zipper for tough, flexible closure
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
Chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist.