missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DuPont™ Tychem™ BR Splash Suit
Lightweight fabric has composite film on one side of a nonwoven substrate
Manufacturer: DuPont™ BR528TYLLG000100
Description
Lightweight, safety-yellow Tychem™ BR fabric is 16mil thick and weighs 6.6 oz. per sq. yd. Offers broad protection for hazardous materials handlers. Tear-, puncture-, and abrasion-resistant.
- Fully encapsulated Level B splash suit
- Large 20mil PVC faceshield
- Attached boots with outer boot flaps
- Expanded back accommodates SCBA
- Elastic wrists
- Two exhaust vents
- Zipper closure with double storm flap
- Taped seams
Chemical and WMD handling, petrochemical manufacturing, emergency response and military applications
Specifications
Attached socks,Boot overflaps | |
Tychem™ BR | |
Elastic | |
Chemical and WMD handling, petrochemical manufacturing, emergency response and military applications | |
1.67 to 1.8 m | |
74.29 cm | |
Large | |
30.75 in. | |
78 cm |
Hook and loop with double storm flap | |
Yellow | |
BR encapsulated level B suit, expanded back, rear entry, standard visor, 1 layer: 20 mil PVC, elastic wrist, attached socks with outer boot flaps, double storm flap with hook and loop closure, two exhaust vents | |
5.5 to 5.9 ft. | |
29.25 in. | |
Taped | |
38.75 to 42.25 in. | |
98.42 to 107.31 cm | |
18 mil |