missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DuPont ™ Tychem ™ BR NFPA 1994 Class 2 Coveralls

An intermediate level of chemical, liquid, and vapor protection designed for situations requiring an SCBA

$795.09 - $842.78

Specifications

Certifications/Compliance NFPA 1992, NFPA 1994, Class 2
Color Bright Yellow
Seam Type Taped
For Use With (Application) Limited-use protective ensemble offers increased vapor and liquid protection without full encapsulation
Thickness 18mil
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Height (English) Height (Metric) Size Price Quantity  
Catalog Number Mfr. No. Height (English) Height (Metric) Size Price Quantity  
19-066-101 Dupont Canada Inc
BR611TYLLG000100
5.5 to 5.9 ft. 1.67 to 1.8m Large Case for $795.09
19-066-100 Dupont Canada Inc
BR611TYLMD000100
5.3 to 5.7 ft. 1.6 to 1.73m Medium Case for $795.09
19-066-102 Dupont Canada Inc
BR611TYLXL000100
5.8 to 6.2 ft. 1.76 to 1.88m X-Large Case for $795.09
19-066-103 Dupont Canada Inc
BR611TYL2X000100
6 to 6.4 ft. 1.82 to 1.95m 2X-Large Case for $842.78
Specifications

Description

Limited-use protective ensemble offers increased vapor and liquid protection without full encapsulation.

  • Tear-, puncture-, and abrasion-resistant fabric offers comfort and flexibility
  • Integrated gloves, face seal, and respirator for liquid integrity
  • Rear-entry zipper closure with a triple storm flap
  • Basis weight: 6.6 oz./sq. yd.
  • Thickness: 16mil
  • High-visibility yellow

Designed to be used with NIOSH approved CBRN SCBA respirators.

Compliance

Meets NFPA 1994 Class 2 standards.

Description & Specifications

Specifications

NFPA 1992, NFPA 1994, Class 2
Taped
18mil
Bright Yellow
Limited-use protective ensemble offers increased vapor and liquid protection without full encapsulation