Learn More
DuPont™ Tychem™ BR Encapsulated Suit
Encapsulated design provides protection from liquid splash exposure for both the wearer and respiratory equipment
Manufacturer: DuPont™ BR528TYL4X0001NY
Description
Tychem BR fabric provides at least 30 minutes of protection against more than 280 chemical challenges, including chemical warfare agents
- Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Integrated socks composed of garment material
- Standard 20 mil PVC faceshield
- Elastic opening for tighter fit at wrist
- Yellow
Model BR527TYL
Front entry design allows wearer to partially don garment (maintain standby position) without assistance
Model BR528TYL
Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
For use in chemical handling, PCB cleanup, hazardous materials/waste cleanup, fire departments, industrial hazmat teams, and utilities
Specifications
Hook and loop | |
Yellow | |
Chemical handling, PCB cleanup, hazardous materials, waste cleanup, fire departments, industrial hazmat teams and utilities | |
1.98 to 2.04 m | |
83.1 cm | |
4X-Large | |
34.25 in. | |
16 mil |
Tychem™ BR | |
Encapsulated level B suit, expanded back, elastic wrist, attached socks with outer boot flaps, double storm flap; Multilayer film barrier laminated to Polypropylene substrate | |
6.5 to 6.7 ft. | |
32.75 in. | |
Taped | |
52.75 to 56.25 in. | |
133.98 to 142.87 cm | |
33 in. |