Please login to your online account to display your discounted pricing
DuPont™ SureStep Series 440 Shoe Covers
Safe, clean and strong shoe cover for controlled environments
$97.10 - $97.10
Specifications
Coating Polyethylene
Closure Type Elastic
Color White
Material Spunbond Polypropylene
Product Type Shoe Covers
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
19-045-452C
dupont canada inc
PE440SWHMD020000
Medium Case of 200 Each for $97.10
Description & Specifications
Specifications
Coating Polyethylene
Closure Type Elastic
Color White
Material Spunbond Polypropylene
Product Type Shoe Covers
Sole Type Non-skid
Height (English) 5.5 in.
Height (Metric) 13.97cm
Seam Type Serged
Generous sizing accommodates all shoe sizes.
  • High slip resistance both wet and dry
  • Spunbounded polypropylene with extruded polyethylene coating
  • Superior liquid resistant fabric
  • Low linting, resists delamination
  • Serged seams, elastic opening and defined toe and heel
  • Available with conductive strip
  • White
Bulk packaged in double transparent poly liners.